missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-nitrobenzoic acid, 98%

Product Code. 11449607
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449607 25 g 25g
11439607 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449607 Supplier Thermo Scientific Alfa Aesar Supplier No. B22100.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7304-32-7 | C7H4FNO4 | 185.11 g/mol

2-Fluoro-5-nitrobenzoic acid may be used in the synthesis of dibenz[b,f]oxazepin-11(10H)-ones, via the nucleophilic aromatic substitution (SNAr) of fluorine in 2-fluoro-5-nitrobenzoic acid with the OH of various 2-aminophenols on solid support, synthesis of substituted dibenzazocines on solid support, via SNAr of fluorine in 2-fluoro-5-nitrobenzoic acid with the OH function of the immobilized polysubstituted phenols, in the synthesis of oxazepines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-5-nitrobenzoic acid may be used in the synthesis of dibenz[b,f]oxazepin-11(10H)-ones, via the nucleophilic aromatic substitution (SNAr) of fluorine in 2-fluoro-5-nitrobenzoic acid with the OH of various 2-aminophenols on solid support, synthesis of substituted dibenzazocines on solid support, via SNAr of fluorine in 2-fluoro-5-nitrobenzoic acid with the OH function of the immobilized polysubstituted phenols, in the synthesis of oxazepines.

Solubility
Soluble in chloroform, ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7304-32-7
Molecular Formula C7H4FNO4
Molecular Weight (g/mol) 185.11
MDL Number MFCD00134238
InChI Key ICXSHFWYCHJILC-UHFFFAOYSA-N
Synonym 2-fluoro-5-nitrobenzoic acid, 2-fluoro-5-nitrobenzoicacid, 2-fluoro-5-nitro-benzoic acid, benzoic acid, 2-fluoro-5-nitro, 2-fluoro-5-nitro-benzoicacid, pubchem1307, acmc-209op6, ksc377e3h, 2-fluoro-5-nitrobenzotc acid, 2-fluoro-5-nitro benzoic acid
PubChem CID 280997
IUPAC Name 2-fluoro-5-nitrobenzoic acid
SMILES OC(=O)C1=CC(=CC=C1F)[N+]([O-])=O

Specifications

Melting Point 142°C to 144°C
Quantity 25 g
Beilstein 1912835
Solubility Information Soluble in chloroform,ethanol.
Formula Weight 185.11
Percent Purity 98%
Chemical Name or Material 2-Fluoro-5-nitrobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.