missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-nitroanisole, TRC

Product Code. 30393868
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30393868 100 mg
30400935 250 mg
30392938 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393868 Supplier Toronto Research Chemicals Supplier No. TRCF597580100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 454-16-0
Molecular Formula C7H6FNO3
Molecular Weight (g/mol) 171.13
SMILES COc1cc(ccc1F)[N+](=O)[O-]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C7H6FNO3/c1-12-7-4-5(9(10)11)2-3-6(7)8/h2-4H,1H3
Formula Weight 171.0332
Chemical Name or Material 2-Fluoro-5-nitroanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.