missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-methylsulphonylnitrobenzene, 95%, Thermo Scientific™

Product Code. 10791452 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10791452 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10791452 Supplier Thermo Scientific Acros Supplier No. 425000025
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 453-72-5
CAS Min % 94.0
CAS Max % 100.0
Melting Point 152°C to 158°C
Color Green-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Molecular Formula C7H6FNO4S
MDL Number MFCD00025066
Quantity 2.5 g
Synonym 4-methylsulfonyl-2-nitrofluorobenzene, 1-fluoro-4-methylsulfonyl-2-nitrobenzene, 1-fluoro-4-methanesulfonyl-2-nitrobenzene, 2-fluoro-5-methyl sulfonyl nitrobenzene, 2-fluoro-5-methylsulphonylnitrobenzene, 4-fluoro-3-nitrophenyl methyl sulfone, benzene,1-fluoro-4-methylsulfonyl-2-nitro, 2-fluoro-5-methylsulfonylnitrobenzene, 2-fluoro-5-methylsulphonyl nitrobenzene, 1-fluoro-4-methylsulfonyl-2-nitro-benzene
InChI Key OUSNDSFSTBZESM-UHFFFAOYSA-N
SMILES CS(=O)(=O)C1=CC=C(F)C(=C1)[N+]([O-])=O
IUPAC Name 1-fluoro-4-methanesulfonyl-2-nitrobenzene
Molecular Weight (g/mol) 219.19
PubChem CID 313137
Formula Weight 219.19
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-5-methylsulphonylnitrobenzene, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Fluoro-5-methylsulphonylnitrobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.