Learn More
2-Fluoro-5-methyl-4-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™
£33.35 - £110.00
Chemical Identifiers
CAS | 1323966-23-9 |
---|---|
Molecular Formula | C9H7BrF4 |
Molecular Weight (g/mol) | 271.053 |
MDL Number | MFCD19687198 |
InChI Key | URFTTWLWYFEASU-UHFFFAOYSA-N |
Synonym | 2-fluoro-5-methyl-4-trifluoromethyl benzyl bromide, 1-bromomethyl-2-fluoro-5-methyl-4-trifluoromethyl benzene |
PubChem CID | 66523487 |
IUPAC Name | 1-(bromomethyl)-2-fluoro-5-methyl-4-(trifluoromethyl)benzene |
SMILES | CC1=CC(=C(C=C1C(F)(F)F)F)CBr |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1323966-23-9 | |
271.053 | |
URFTTWLWYFEASU-UHFFFAOYSA-N | |
66523487 | |
CC1=CC(=C(C=C1C(F)(F)F)F)CBr |
C9H7BrF4 | |
MFCD19687198 | |
2-fluoro-5-methyl-4-trifluoromethyl benzyl bromide, 1-bromomethyl-2-fluoro-5-methyl-4-trifluoromethyl benzene | |
1-(bromomethyl)-2-fluoro-5-methyl-4-(trifluoromethyl)benzene |
Specifications
1323966-23-9 | |
MFCD19687198 | |
Moisture sensitive | |
URFTTWLWYFEASU-UHFFFAOYSA-N | |
1-(bromomethyl)-2-fluoro-5-methyl-4-(trifluoromethyl)benzene | |
66523487 | |
97% | |
2-Fluoro-5-methyl-4-(trifluoromethyl)benzyl bromide |
C9H7BrF4 | |
UN3265 | |
2-fluoro-5-methyl-4-trifluoromethyl benzyl bromide, 1-bromomethyl-2-fluoro-5-methyl-4-trifluoromethyl benzene | |
CC1=CC(=C(C=C1C(F)(F)F)F)CBr | |
271.053 | |
271.05 | |
Liquid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures