missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-methyl-4-(trifluoromethyl)benzamide, 97%, Thermo Scientific™

Catalog Number 15478715
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1323966-43-3
C9H7F4NO
221.155
MFCD19687194
YHTYNOXDHGBXMS-UHFFFAOYSA-N
2-fluoro-5-methyl-4-trifluoromethyl benzamide
66523632
2-fluoro-5-methyl-4-(trifluoromethyl)benzamide
CC1=C(C=C(C(=C1)C(=O)N)F)C(F)(F)F
This item is not returnable. View return policy
1323966-43-3
C9H7F4NO
221.155
MFCD19687194
YHTYNOXDHGBXMS-UHFFFAOYSA-N
2-fluoro-5-methyl-4-trifluoromethyl benzamide
66523632
2-fluoro-5-methyl-4-(trifluoromethyl)benzamide
CC1=C(C=C(C(=C1)C(=O)N)F)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 221.15
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Fluoro-5-methyl-4-(trifluoromethyl)benzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'