missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-methyl-4-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™

Product Code. p-7052486
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15146695 1 g 1g
15418815 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 15146695

Brand: Thermo Scientific Alfa Aesar H3417403

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1323966-40-0
Molecular Formula C9H6F4O
Molecular Weight (g/mol) 206.14
MDL Number MFCD19687179
InChI Key GKMVOIBAXNLEOE-UHFFFAOYSA-N
Synonym 2-fluoro-5-methyl-4-trifluoromethyl benzaldehyde
PubChem CID 66523595
IUPAC Name 2-fluoro-5-methyl-4-(trifluoromethyl)benzaldehyde
SMILES CC1=C(C=C(C(=C1)C=O)F)C(F)(F)F

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 206.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Fluoro-5-methyl-4-(trifluoromethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.