missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-methyl-4-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™

Product Code. 15418815
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1323966-40-0
Molecular Formula C9H6F4O
Molecular Weight (g/mol) 206.14
MDL Number MFCD19687179
InChI Key GKMVOIBAXNLEOE-UHFFFAOYSA-N
Synonym 2-fluoro-5-methyl-4-trifluoromethyl benzaldehyde
PubChem CID 66523595
IUPAC Name 2-fluoro-5-methyl-4-(trifluoromethyl)benzaldehyde
SMILES CC1=C(C=C(C(=C1)C=O)F)C(F)(F)F

Specifications

Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 206.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Fluoro-5-methyl-4-(trifluoromethyl)benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'