missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(methoxycarbonylmethyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Product Code. 15427846
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15427846

Brand: Thermo Scientific Alfa Aesar H62786.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 944317-66-2
Molecular Formula C15H20BFO4
Molecular Weight (g/mol) 294.13
MDL Number MFCD12026092
InChI Key LTWSFTBHFDGCTE-UHFFFAOYSA-N
PubChem CID 46739456
IUPAC Name methyl 2-[4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate
SMILES COC(=O)CC1=CC(B2OC(C)(C)C(C)(C)O2)=C(F)C=C1

Specifications

Quantity 250 mg
Synonym methyl 2-4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, 2-fluoro-5-methoxycarbonylmethyl phenylboronic acid, pinacol ester, methyl 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, 2-fluoro-5-methoxycarbonylmethyl phenylboronic acid,pinacol ester, 2-fluoro-5-methoxycarbonylmethyl benzeneboronic acid pinacol ester, methyl 2-4-fluoro-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, methyl2-4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-acetic acid methyl ester
Formula Weight 294.13
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-Fluoro-5-(methoxycarbonylmethyl)benzeneboronic acid pinacol ester
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.