missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-methoxybenzonitrile, 98%, Thermo Scientific Chemicals

Product Code. 15475477
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15475477 5 g 5g
15465477 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15475477 Supplier Thermo Scientific Alfa Aesar Supplier No. H32724.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127667-01-0
Molecular Formula C8H6FNO
Molecular Weight (g/mol) 151.14
MDL Number MFCD00142921
InChI Key VBZLRHYLNXWZIU-UHFFFAOYSA-N
Synonym 2-fluoro-5-methoxy-benzonitrile, benzonitrile, 2-fluoro-5-methoxy, 3-cyano-4-fluoroanisole, fluoromethoxybenzonitrile2, 2-fluoro-5-methoxybenzenecarbonitrile, pubchem3500, acmc-209bct, ksc494m9f, 2-fluoro-5-methoxy benzonitrile, buttpark 80\01-21
PubChem CID 2774546
IUPAC Name 2-fluoro-5-methoxybenzonitrile
SMILES COC1=CC(C#N)=C(F)C=C1

Specifications

Melting Point 81°C to 85°C
Quantity 5 g
UN Number UN3439
Formula Weight 151.14
Percent Purity 98%
Chemical Name or Material 2-Fluoro-5-methoxybenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.