missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-methoxybenzaldehyde, 97%

Product Code. 11493148
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493148 25 g 25g
11473148 1 g 1g
11483148 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493148 Supplier Thermo Scientific Alfa Aesar Supplier No. A18904.14
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 105728-90-3 | C8H7FO2 | 154.14 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105728-90-3
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00070795
InChI Key DKIQXHIAEMGZGO-UHFFFAOYSA-N
Synonym 6-fluoro-m-anisaldehyde, 2-fluoro-5-methoxy-benzaldehyde, 2-fluoro-5-methoxy benzaldehyde, benzaldehyde, 2-fluoro-5-methoxy, pubchem2634, 4-fluoro-3-formylanisole, acmc-2098hi, ksc490m2b, 2-fluoro-5-methyloxy benzaldehyde, 2-fluoro-5-methoxybenzaldehyde
PubChem CID 2734872
IUPAC Name 2-fluoro-5-methoxybenzaldehyde
SMILES COC1=CC(=C(C=C1)F)C=O

Specifications

Density 1.237
Boiling Point 80°C (0.9 mmHg)
Flash Point 107°C (224°F)
Refractive Index 1.533
Quantity 25 g
Beilstein 4379088
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 154.14
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-methoxybenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.