missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-methoxy-3-(trifluoromethyl)benzyl alcohol, 97%, Thermo Scientific™

Product Code. 15408585
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15408585 1 g 1g
15418585 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15408585 Supplier Thermo Scientific Alfa Aesar Supplier No. H33682.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1373921-03-9
Molecular Formula C9H8F4O2
Molecular Weight (g/mol) 224.155
MDL Number MFCD22201037
InChI Key ZWHWVTLVMXSITQ-UHFFFAOYSA-N
Synonym 2-fluoro-5-methoxy-3-trifluoromethyl benzyl alcohol, 2-fluoro-5-methoxy-3-trifluoromethyl phenyl methanol
PubChem CID 86277639
IUPAC Name [2-fluoro-5-methoxy-3-(trifluoromethyl)phenyl]methanol
SMILES COC1=CC(=C(C(=C1)CO)F)C(F)(F)F

Specifications

Quantity 1 g
Formula Weight 224.15
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-methoxy-3-(trifluoromethyl)benzyl alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.