missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(4-oxo-3,4-dihydrophthalazin-1-ylmethyl)benzoic Acid, TRC

Product Code. 17982645
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17982645 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17982645 Supplier Toronto Research Chemicals Supplier No. TRCF593022250MG
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 763114-26-7
Molecular Formula C16 H11 F N2 O3
Quantity 250 mg
Synonym 5-[(3,4-Dihydro-4-oxo-1-phthalazinyl)methyl]-2-fluorobenzoic Acid, 2-Fluoro-5-[(4-oxo-3H-phthalazin-1-yl)methyl]benzoic acid, Benzoic acid, 5-[(3,4-dihydro-4-oxo-1-phthalazinyl)methyl]-2-fluoro- (9CI, ACI), 5-[(3,4-Dihydro-4-oxo-1-phthalazinyl)methyl]-2-fluorobenzoic acid (ACI), 2-Fluoro-5-(4-oxo-3,4-dihydrophthalazin-1-ylmethyl)benzoic acid, 2-Fluoro-5-[(4-oxo-3H-phthalazin-1-yl)methyl]benzoic acid
InChI Formula InChI=1S/C16H11FN2O3/c17-13-6-5-9(7-12(13)16(21)22)8-14-10-3-1-2-4-11(10)15(20)19-18-14/h1-7H,8H2,(H,19,20)(H,21,22)
SMILES OC(=O)c1cc(CC2=NNC(=O)c3ccccc23)ccc1F
IUPAC Name 2-fluoro-5-[(4-oxo-3H-phthalazin-1-yl)methyl]benzoic acid
Molecular Weight (g/mol) 298.27
Formula Weight 298.0754
Chemical Name or Material 2-Fluoro-5-(4-oxo-3,4-dihydrophthalazin-1-ylmethyl)benzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.