missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(3-pyridylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific™

£87.55 - £271.00

Chemical Identifiers

CAS 1449132-32-4
Molecular Formula C12H10BFN2O3
Molecular Weight (g/mol) 260.031
MDL Number MFCD20265175
InChI Key LWVDXBKALYJJIU-UHFFFAOYSA-N
Synonym 2-fluoro-5-3-pyridylcarbamoyl benzeneboronic acid, 2-fluoro-5-pyridin-3-yl carbamoyl phenylboronic acid, 2-fluoro-5-3-pyridylcarbamoyl phenylboronic acid
PubChem CID 73995628
IUPAC Name [2-fluoro-5-(pyridin-3-ylcarbamoyl)phenyl]boronic acid
SMILES B(C1=C(C=CC(=C1)C(=O)NC2=CN=CC=C2)F)(O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15480756
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H54093.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15470756
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H54093.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1449132-32-4
260.031
LWVDXBKALYJJIU-UHFFFAOYSA-N
73995628
B(C1=C(C=CC(=C1)C(=O)NC2=CN=CC=C2)F)(O)O
C12H10BFN2O3
MFCD20265175
2-fluoro-5-3-pyridylcarbamoyl benzeneboronic acid, 2-fluoro-5-pyridin-3-yl carbamoyl phenylboronic acid, 2-fluoro-5-3-pyridylcarbamoyl phenylboronic acid
[2-fluoro-5-(pyridin-3-ylcarbamoyl)phenyl]boronic acid

Specifications

1449132-32-4
MFCD20265175
LWVDXBKALYJJIU-UHFFFAOYSA-N
[2-fluoro-5-(pyridin-3-ylcarbamoyl)phenyl]boronic acid
73995628
97%
C12H10BFN2O3
2-fluoro-5-3-pyridylcarbamoyl benzeneboronic acid, 2-fluoro-5-pyridin-3-yl carbamoyl phenylboronic acid, 2-fluoro-5-3-pyridylcarbamoyl phenylboronic acid
B(C1=C(C=CC(=C1)C(=O)NC2=CN=CC=C2)F)(O)O
260.031
260.03
2-Fluoro-5-(3-pyridylcarbamoyl)benzeneboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications