missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals

Product Code. 15465987
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15465987 5 g 5g
15455987 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15465987 Supplier Thermo Scientific Alfa Aesar Supplier No. H33198.06
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 118078-66-3 | Molecular Formula: C6H3F4N | Molecular Weight: 165.091 g/mol

2-Fluoro-4-(trifluoromethyl)pyridine acts as a reactant in the preparation of aminopyridines through amination reactions. It is also used as a catalytic ligand for regioselective preparation of tetramethylbiphenyls through aerobic oxidative coupling of xylene catalyzed by palladium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-4-(trifluoromethyl)pyridine acts as a reactant in the preparation of aminopyridines through amination reactions. It is also used as a catalytic ligand for regioselective preparation of tetramethylbiphenyls through aerobic oxidative coupling of xylene catalyzed by palladium.

Solubility
Miscible with water, alcohol, acetonitrile and N,N-dimethylformamide

Notes
Incompatible with oxidizing agents and bases.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118078-66-3
Molecular Formula C6H3F4N
Molecular Weight (g/mol) 165.091
MDL Number MFCD09879262
InChI Key DFNQBXZKPUBEIX-UHFFFAOYSA-N
Synonym 2-fluoro-4-trifluoromethyl pyridine, 2-fluoro-4-trifluoromethyl-pyridine, pyridine, 2-fluoro-4-trifluoromethyl, pubchem23563, acmc-1c7kk, 2-fluoranyl-4-trifluoromethyl pyridine
PubChem CID 11159514
IUPAC Name 2-fluoro-4-(trifluoromethyl)pyridine
SMILES C1=CN=C(C=C1C(F)(F)F)F

Specifications

Boiling Point 113°C
Quantity 5 g
Solubility Information Miscible with water,alcohol,acetonitrile and N,N-dimethylformamide
Formula Weight 165.09
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.