missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(trifluoromethyl)pyridine-3-carboxylic acid, Maybridge

Product Code. 10607200
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10607200

Brand: Thermo Scientific Maybridge MAY00203DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1040681-74-0
Packaging Amber Glass Bottle
Molecular Formula C7H3F4NO2
Quantity 1 g
Synonym 2-fluoro-4-trifluoromethyl pyridine-3-carboxylic acid, 2-fluoro-4-trifluoromethyl pyridine-3-carboxylicacid, 2-fluoro-4-trifluoromethyl nicotinic acid, acmc-20anv9
InChI Key ABXMDMDQUVWALB-UHFFFAOYSA-N
SMILES C1=CN=C(C(=C1C(F)(F)F)C(=O)O)F
IUPAC Name 2-fluoro-4-(trifluoromethyl)pyridine-3-carboxylic acid
Molecular Weight (g/mol) 209.1
PubChem CID 28307766
Formula Weight 209.1
Percent Purity 97%
Chemical Name or Material 2-fluoro-4-(trifluoromethyl)pyridine-3-carboxylic acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.