missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(trifluoromethyl)nicotinic acid, 97%

Product Code. 15498135
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15498135

Brand: Thermo Scientific Alfa Aesar H32618.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1040681-74-0 | C7H3F4NO2 | 209.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1040681-74-0
Molecular Formula C7H3F4NO2
Molecular Weight (g/mol) 209.1
MDL Number MFCD10687403
InChI Key ABXMDMDQUVWALB-UHFFFAOYSA-N
Synonym 2-fluoro-4-trifluoromethyl pyridine-3-carboxylic acid, 2-fluoro-4-trifluoromethyl pyridine-3-carboxylicacid, 2-fluoro-4-trifluoromethyl nicotinic acid, acmc-20anv9
PubChem CID 28307766
IUPAC Name 2-fluoro-4-(trifluoromethyl)pyridine-3-carboxylic acid
SMILES C1=CN=C(C(=C1C(F)(F)F)C(=O)O)F

Specifications

Melting Point 91°C to 93°C
Quantity 250 mg
Formula Weight 209.1
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)nicotinic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.