missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(trifluoromethoxy)benzamide, 97%, Thermo Scientific™

Catalog Number 15487167
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
1240257-18-4
C8H5F4NO2
223.127
MFCD16652438
XIMSWKYWHVBWDF-UHFFFAOYSA-N
2-fluoro-4-trifluoromethoxy benzamide
66523444
2-fluoro-4-(trifluoromethoxy)benzamide
C1=CC(=C(C=C1OC(F)(F)F)F)C(=O)N
This item is not returnable. View return policy
1240257-18-4
C8H5F4NO2
223.127
MFCD16652438
XIMSWKYWHVBWDF-UHFFFAOYSA-N
2-fluoro-4-trifluoromethoxy benzamide
66523444
2-fluoro-4-(trifluoromethoxy)benzamide
C1=CC(=C(C=C1OC(F)(F)F)F)C(=O)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 223.12
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-(trifluoromethoxy)benzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'