missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(trifluoromethoxy)benzaldehyde, 95%

Product Code. 15455997
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15455997 5 g 5g
15445997 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15455997 Supplier Thermo Scientific Alfa Aesar Supplier No. H33203.06
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1227628-83-2 | C8H4F4O2 | 208.112 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1227628-83-2
Molecular Formula C8H4F4O2
Molecular Weight (g/mol) 208.112
MDL Number MFCD16652440
InChI Key DEACWPDDLIOZJV-UHFFFAOYSA-N
Synonym 2-fluoro-4-trifluoromethoxy benzaldehyde
PubChem CID 53465593
IUPAC Name 2-fluoro-4-(trifluoromethoxy)benzaldehyde
SMILES C1=CC(=C(C=C1OC(F)(F)F)F)C=O

Specifications

Refractive Index 1.4455
Quantity 5 g
Sensitivity Air sensitive
Formula Weight 208.11
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-(trifluoromethoxy)benzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.