missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(trifluoromethoxy)benzaldehyde, 95%

Product Code. 15445997
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15445997 1 g 1g
15455997 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 15445997

Brand: Thermo Scientific Alfa Aesar H33203.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1227628-83-2 | C8H4F4O2 | 208.112 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1227628-83-2
Molecular Formula C8H4F4O2
Molecular Weight (g/mol) 208.112
MDL Number MFCD16652440
InChI Key DEACWPDDLIOZJV-UHFFFAOYSA-N
Synonym 2-fluoro-4-trifluoromethoxy benzaldehyde
PubChem CID 53465593
IUPAC Name 2-fluoro-4-(trifluoromethoxy)benzaldehyde
SMILES C1=CC(=C(C=C1OC(F)(F)F)F)C=O

Specifications

Refractive Index 1.4455
Quantity 1 g
Sensitivity Air sensitive
Formula Weight 208.11
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-(trifluoromethoxy)benzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.