missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(pentafluorothio)phenylacetic acid, 97%, Thermo Scientific™

Catalog Number 15428665
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1240257-93-5
C8H6F6O2S
280.184
MFCD16652509
ZTDHXJDCNBDLPP-UHFFFAOYSA-N
2-fluoro-4-pentafluorosulfur phenylacetic acid, 2-fluoro-4-pentafluorothio phenylacetic acid, 2-fluoro-4-pentafluoro-??-sulfanyl phenyl acetic acid
66523510
2-[2-fluoro-4-(pentafluoro-$l^{6}-sulfanyl)phenyl]acetic acid
C1=CC(=C(C=C1S(F)(F)(F)(F)F)F)CC(=O)O
This item is not returnable. View return policy
1240257-93-5
C8H6F6O2S
280.184
MFCD16652509
ZTDHXJDCNBDLPP-UHFFFAOYSA-N
2-fluoro-4-pentafluorosulfur phenylacetic acid, 2-fluoro-4-pentafluorothio phenylacetic acid, 2-fluoro-4-pentafluoro-??-sulfanyl phenyl acetic acid
66523510
2-[2-fluoro-4-(pentafluoro-$l^{6}-sulfanyl)phenyl]acetic acid
C1=CC(=C(C=C1S(F)(F)(F)(F)F)F)CC(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 280.18
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-(pentafluorothio)phenylacetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'