missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-nitroaniline, 98%, Thermo Scientific™

Product Code. 15585256 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15585256 100 g 100g
15565256 5 g 5g
15575256 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15585256 Supplier Thermo Scientific Acros Supplier No. 457601000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 369-35-7
Molecular Formula C6H5FN2O2
Molecular Weight (g/mol) 156.11
InChI Key LETNCFZQCNCACQ-UHFFFAOYSA-N
Synonym 2-fluoro-4-nitrobenzenamine, 2-fluoro-4-nitro-phenylamine, benzenamine, 2-fluoro-4-nitro, 2-fluoro-4-nitrophenylamine, 4-amino-3-fluoronitrobenzene, 2-fluoro-4-nitroaniline, pubchem8503, acmc-1bncp, 2-fluoro-4-niroaniline, 2-fluoro-4-nitro-aniline
PubChem CID 101254
IUPAC Name 2-fluoro-4-nitroaniline
SMILES C1=CC(=C(C=C1[N+](=O)[O-])F)N

Specifications

Melting Point 122.0°C to 130.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Linear Formula FC6H3(NO2)NH2
Quantity 100 g
Formula Weight 156.11
Percent Purity 98%
Chemical Name or Material 2-Fluoro-4-nitroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.