Learn More
Alfa Aesar™ 2-Fluoro-4-nitro-N,N-di-n-propylaniline, 97%
Brand: Alfa Aesar™ H63332.06
Code : NEW
Additional Details : Weight : 0.00500kg Transport : UN number : 2811 Chem class : 1 Pack group : III
Quantity | 5g |
---|
Chemical Identifiers
2-Fluoro-4-nitro-N,N-di-n-propylaniline | |
240.27 | |
UN2811 | |
(2-Fluoro-4-nitrophenyl)di-n-propylamine |
Specifications
2-Fluoro-4-nitro-N,N-di-n-propylaniline | |
Solid | |
240.27 | |
(2-Fluoro-4-nitrophenyl)di-n-propylamine |
5g | |
97% | |
UN2811 |