missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(methylsulfonyl)benzoic Acid, TRC

Product Code. 30399819
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399819 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399819 Supplier Toronto Research Chemicals Supplier No. TRCF596893500MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material 2-Fluoro-4-(methylsulfonyl)benzoic Acid
CAS 142994-04-5
Molecular Formula C12 H14 F3 N O
Quantity 500 mg
Synonym 2-Fluoro-4-mesylbenzoic Acid
InChI Formula InChI=1S/C12H14F3NO/c13-12(14,15)10-3-1-2-9(8-10)11(17)4-6-16-7-5-11/h1-3,8,16-17H,4-7H2
SMILES OC1(CCNCC1)c2cccc(c2)C(F)(F)F
IUPAC Name 4-[3-(trifluoromethyl)phenyl]piperidin-4-ol
Molecular Weight (g/mol) 245.241
Formula Weight 245.103
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.