missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-methylphenylboronic Acid, TRC

Product Code. 30392300
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392300 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392300 Supplier Toronto Research Chemicals Supplier No. TRCF5937501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 170981-26-7
Molecular Formula C7 H8 B F O2
Quantity 1 g
InChI Formula InChI=1S/C7H8BFO2/c1-5-2-3-6(8(10)11)7(9)4-5/h2-4,10-11H,1H3
SMILES Cc1ccc(B(O)O)c(F)c1
IUPAC Name (2-fluoro-4-methylphenyl)boronic acid
Molecular Weight (g/mol) 153.95
Formula Weight 154.0601
Chemical Name or Material 2-Fluoro-4-methylphenylboronic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.