missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-methylbenzyl bromide, 97%, Thermo Scientific™

Product Code. 11481410
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11481410 1 g 1g
11491410 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11481410 Supplier Thermo Scientific Alfa Aesar Supplier No. H26440.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Fluoro-4-methylbenzyl bromide is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118745-63-4
Molecular Formula C8H8BrF
Molecular Weight (g/mol) 203.05
MDL Number MFCD03094327
InChI Key SSDOQXRIONFILC-UHFFFAOYSA-N
PubChem CID 2778475
IUPAC Name 1-(bromomethyl)-2-fluoro-4-methylbenzene
SMILES CC1=CC(F)=C(CBr)C=C1

Specifications

Refractive Index 1.5465
Quantity 1 g
UN Number UN3265
Synonym 2-fluoro-4-methylbenzyl bromide, 1-bromomethyl-2-fluoro-4-methylbenzene, 1-bromomethyl-2-fluoro-4-methyl-benzene, acmc-1c5r8, benzene,1-bromomethyl-2-fluoro-4-methyl, benzene, 1-bromomethyl-2-fluoro-4-methyl
Solubility Information Insoluble in water.
Formula Weight 203.06
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-methylbenzyl bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.