Learn More
2-Fluoro-4-methylbenzoyl chloride, 97%, Thermo Scientific™
£55.86 - £220.00
Chemical Identifiers
CAS | 59189-98-9 |
---|---|
Molecular Formula | C8H6ClFO |
Molecular Weight (g/mol) | 172.58 |
MDL Number | MFCD04115879 |
InChI Key | SVSVOPWCUHQFAM-UHFFFAOYSA-N |
Synonym | benzoyl chloride, 2-fluoro-4-methyl-9ci, 2-fluoro-4-methyl-benzoyl chloride, benzoyl chloride, 2-fluoro-4-methyl |
PubChem CID | 5173367 |
IUPAC Name | 2-fluoro-4-methylbenzoyl chloride |
SMILES | CC1=CC(F)=C(C=C1)C(Cl)=O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is employed as a intermediates for pharmaceutical.
Solubility
Reacts with water.
Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
Chemical Identifiers
59189-98-9 | |
172.58 | |
SVSVOPWCUHQFAM-UHFFFAOYSA-N | |
5173367 | |
CC1=CC(F)=C(C=C1)C(Cl)=O |
C8H6ClFO | |
MFCD04115879 | |
benzoyl chloride, 2-fluoro-4-methyl-9ci, 2-fluoro-4-methyl-benzoyl chloride, benzoyl chloride, 2-fluoro-4-methyl | |
2-fluoro-4-methylbenzoyl chloride |
Specifications
59189-98-9 | |
C8H6ClFO | |
UN3261 | |
benzoyl chloride, 2-fluoro-4-methyl-9ci, 2-fluoro-4-methyl-benzoyl chloride, benzoyl chloride, 2-fluoro-4-methyl | |
SVSVOPWCUHQFAM-UHFFFAOYSA-N | |
2-fluoro-4-methylbenzoyl chloride | |
5173367 | |
97% |
43°C to 47°C | |
MFCD04115879 | |
Moisture sensitive | |
Reacts with water. | |
CC1=CC(F)=C(C=C1)C(Cl)=O | |
172.58 | |
172.59 | |
2-Fluoro-4-methylbenzoyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures