missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-methoxybenzonitrile, 97%, Thermo Scientific™

Product Code. 15415267
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15415267 5 g 5g
15405267 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15415267 Supplier Thermo Scientific Alfa Aesar Supplier No. H32531.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Fluoro-4-methoxybenzonitrile is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94610-82-9
Molecular Formula C8H6FNO
Molecular Weight (g/mol) 151.14
MDL Number MFCD00509374
InChI Key HWKUZTFIZATJPM-UHFFFAOYSA-N
Synonym 3-fluoro-4-cyanoanisole, 2-fluoro-4-methoxy-benzonitrile, benzonitrile, 2-fluoro-4-methoxy, 3-fluoro-4-cyano anisole, 4-cyano-3-fluoroanisole, buttpark 81\01-98, 2-fluoro-4-methoxybenzenecarbonitrile, pubchem4783, 2-fluoro-4-methoxybe
PubChem CID 639878
IUPAC Name 2-fluoro-4-methoxybenzonitrile
SMILES COC1=CC(F)=C(C=C1)C#N

Specifications

Melting Point 62°C to 65°C
Quantity 5 g
UN Number UN3439
Formula Weight 151.14
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-methoxybenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.