missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-methoxybenzoic acid, 95%, Thermo Scientific™

Product Code. 11451390
Click to view available options
Quantity:
500 mg
2.5 g
Unit Size:
2.5g
500mg
This item is not returnable. View return policy

Product Code. 11451390

Brand: Thermo Scientific Alfa Aesar H26426.27

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 394-42-3
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.139
MDL Number MFCD00509373
InChI Key UPWMPIKNUXTWFP-UHFFFAOYSA-N
PubChem CID 2774542
IUPAC Name 2-fluoro-4-methoxybenzoic acid
SMILES COC1=CC(=C(C=C1)C(=O)O)F

Specifications

Melting Point 192°C to 198°C
Quantity 2.5 g
Synonym 2-fluoro-p-anisic acid, 2-fluoro-4-methoxy-benzoic acid, 2-fluoro-4-methoxybenzoicacid, benzoic acid, 2-fluoro-4-methoxy, acmc-1ajdw, 4-carboxy-3-fluoroanisole, ksc494q6n, akos brn-0650, attercop-chm at143177, benzoic acid,2-fluoro-4-methoxy
Formula Weight 170.14
Percent Purity 95%
Chemical Name or Material 2-Fluoro-4-methoxybenzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.