missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-iodopyridine, 97%, Thermo Scientific™

Product Code. 10306674
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10306674 5 g Amber Glass Bottle 5g
10779405 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10306674 Supplier Thermo Scientific Maybridge Supplier No. MO07994DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 22282-70-8
Molecular Formula C5H3FIN
Molecular Weight (g/mol) 222.989
MDL Number MFCD03092926
InChI Key ADPRIAVYIGHFSO-UHFFFAOYSA-N
Synonym 4-iodo-2-fluoropyridine, 2-fluoro-4-iodo-pyridine, 2-fluoro-4-iodo pyridine, pyridine, 2-fluoro-4-iodo, pubchem1231, pubchem3510, fluoro-4-iodo pyridine, acmc-1cg64, ksc201s6h
PubChem CID 7023570
IUPAC Name 2-fluoro-4-iodopyridine
SMILES C1=CN=C(C=C1I)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 222.98
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-iodopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.