missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-iodopyridine, 95%, Thermo Scientific™

Product Code. 11451780
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11451780 5 g 5g
11441780 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11451780 Supplier Thermo Scientific Alfa Aesar Supplier No. H27517.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22282-70-8
Molecular Formula C5H3FIN
Molecular Weight (g/mol) 222.989
MDL Number MFCD03092926
InChI Key ADPRIAVYIGHFSO-UHFFFAOYSA-N
Synonym 4-iodo-2-fluoropyridine, 2-fluoro-4-iodo-pyridine, 2-fluoro-4-iodo pyridine, pyridine, 2-fluoro-4-iodo, pubchem1231, pubchem3510, fluoro-4-iodo pyridine, acmc-1cg64, ksc201s6h
PubChem CID 7023570
IUPAC Name 2-fluoro-4-iodopyridine
SMILES C1=CN=C(C=C1I)F

Specifications

Melting Point 57°C to 58°C
Quantity 5 g
Sensitivity Light sensitive
Formula Weight 222.99
Percent Purity 95%
Chemical Name or Material 2-Fluoro-4-iodopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.