missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-iodobenzonitrile, 98%

Product Code. 15458568
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15458568 5 g 5g
15468568 250 mg 250mg
15448568 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15458568 Supplier Thermo Scientific Alfa Aesar Supplier No. H61973.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 137553-42-5 | C7H3FIN | 247.011 g/mol

Used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137553-42-5
Molecular Formula C7H3FIN
Molecular Weight (g/mol) 247.011
MDL Number MFCD03094172
InChI Key WHQVXHBSTRFRCE-UHFFFAOYSA-N
Synonym 3-fluoro-4-cyano-1-iodobenzene, benzonitrile, 2-fluoro-4-iodo, 2-fluoro-4-iodobenzenecarbonitrile, pubchem4778, acmc-209xcp, intermediates-zcf02061, 4-iodo-2-fluorobenzonitrile, 2-fluoro-4-iodo-benzonitrile, ksc493i5t, benzonitrile,2-fluoro-4-iodo
PubChem CID 2774518
IUPAC Name 2-fluoro-4-iodobenzonitrile
SMILES C1=CC(=C(C=C1I)F)C#N

Specifications

Color Red to Brown
Quantity 5 g
UN Number UN3439
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 247.01
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Fluoro-4-iodobenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.