missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-iodoaniline, 99%

Product Code. 11490607
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490607 5 g 5g
11400617 25 g 25g
11410617 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490607 Supplier Thermo Scientific Alfa Aesar Supplier No. B22450.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29632-74-4 | C6H5FIN | 237.016 g/mol

2-Fluoro-4-iodoaniline is widely used in the pharmaceutical, chemical and food industries as a fundamental building block to create a variety of important compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-4-iodoaniline is widely used in the pharmaceutical, chemical and food industries as a fundamental building block to create a variety of important compounds.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29632-74-4
Molecular Formula C6H5FIN
Molecular Weight (g/mol) 237.016
MDL Number MFCD00011738
InChI Key CUMTUBVTKOYYOU-UHFFFAOYSA-N
Synonym 2-fluoro-4-iodo-phenylamine, benzenamine, 2-fluoro-4-iodo, 2-fluoro-4-iodobenzenamine, 2-fluoro-4-iodo aniline, 2-fluoro-4-iodo-aniline, 2-fluoro-4-iodophenylamine, zlchem 543, pubchem1532, 2-fluoro4-iodoaniline, 2-fluoro-4-iodaniline
PubChem CID 185694
IUPAC Name 2-fluoro-4-iodoaniline
SMILES C1=CC(=C(C=C1I)F)N

Specifications

Melting Point 55°C to 57°C
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 2081100
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 237.02
Percent Purity 99%
Chemical Name or Material 2-Fluoro-4-iodoaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.