missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-iodoaniline, 98%, Thermo Scientific™

Product Code. 10672345 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10672345 5 g Glass bottle 5g
10372823 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10672345 Supplier Thermo Scientific Acros Supplier No. 311930050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 29632-74-4
Molecular Formula C6H5FIN
Molecular Weight (g/mol) 237.01
MDL Number MFCD00011738
InChI Key CUMTUBVTKOYYOU-UHFFFAOYSA-N
Synonym 2-fluoro-4-iodo-phenylamine, benzenamine, 2-fluoro-4-iodo, 2-fluoro-4-iodobenzenamine, 2-fluoro-4-iodo aniline, 2-fluoro-4-iodo-aniline, 2-fluoro-4-iodophenylamine, zlchem 543, pubchem1532, 2-fluoro4-iodoaniline, 2-fluoro-4-iodaniline
PubChem CID 185694
IUPAC Name 2-fluoro-4-iodoaniline
SMILES C1=CC(=C(C=C1I)F)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 51°C to 57°C
Color White to Brown
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula IC6H3(F)NH2
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 237.01
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Fluoro-4-iodoaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.