missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-iodo-5-methylpyridine, 95%, Thermo Scientific™

Catalog Number 15422429
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
153034-94-7
C6H5FIN
237.016
MFCD03095297
BVKQLNXPPQEELX-UHFFFAOYSA-N
2-fluoro-4-iodo-5-picoline, 2-fluoro-4-iodo-5-methyl-pyridine, 6-fluoro-4-iodo-b-picoline, 6-fluoro-4-iodo-3-picoline, 2-fluoro-4-iodo-5-methyl pyridine, pyridine,2-fluoro-4-iodo-5-methyl, pyridine, 2-fluoro-4-iodo-5-methyl, pubchem1305, acmc-20el7f, abbypharma ap-30-1289
10060052
2-fluoro-4-iodo-5-methylpyridine
CC1=CN=C(C=C1I)F
This item is not returnable. View return policy
153034-94-7
C6H5FIN
237.016
MFCD03095297
BVKQLNXPPQEELX-UHFFFAOYSA-N
2-fluoro-4-iodo-5-picoline, 2-fluoro-4-iodo-5-methyl-pyridine, 6-fluoro-4-iodo-b-picoline, 6-fluoro-4-iodo-3-picoline, 2-fluoro-4-iodo-5-methyl pyridine, pyridine,2-fluoro-4-iodo-5-methyl, pyridine, 2-fluoro-4-iodo-5-methyl, pubchem1305, acmc-20el7f, abbypharma ap-30-1289
10060052
2-fluoro-4-iodo-5-methylpyridine
CC1=CN=C(C=C1I)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 40°C to 45°C
Color White
Quantity 250 mg
Beilstein 6643371
Sensitivity Light sensitive
Formula Weight 237.02
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Fluoro-4-iodo-5-methylpyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'