missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-hydroxybenzonitrile, 99%

Product Code. 11407114
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11407114 1 g 1g
11417114 5 g 5g
11427114 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11407114 Supplier Thermo Scientific Alfa Aesar Supplier No. B21535.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 82380-18-5 | C7H4FNO | 137.113 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82380-18-5
Molecular Formula C7H4FNO
Molecular Weight (g/mol) 137.113
MDL Number MFCD00192177
InChI Key REIVHYDACHXPNH-UHFFFAOYSA-N
Synonym 4-cyano-3-fluorophenol, 3-fluoro-4-cyanophenol, 2-fluoro-4-benzonitrile, benzonitrile, 2-fluoro-4-hydroxy, 4-hydroxy-2-fluorobenzonitrile, 2-fluoro-4-hydroxy-benzonitrile, 2-fluoro-4-hydroxybenzenecarbonitrile, pubchem2610, pubchem3495
PubChem CID 2734675
IUPAC Name 2-fluoro-4-hydroxybenzonitrile
SMILES C1=CC(=C(C=C1O)F)C#N

Specifications

Melting Point 123°C to 126°C
Density 1.45
Quantity 1 g
UN Number UN3077
Beilstein 4741066
Formula Weight 137.11
Percent Purity 99%
Chemical Name or Material 2-Fluoro-4-hydroxybenzonitrile
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Fluoro-4-hydroxybenzonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 2-Fluoro-4-hydroxybenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.