missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-hydroxybenzoic acid, 98%, Thermo Scientific™

Product Code. 15555256 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15555256 25 g 25g
15581832 1 g 1g
15545256 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15555256 Supplier Thermo Scientific Acros Supplier No. 457590250
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 65145-13-3
Molecular Formula C7H5FO3
Molecular Weight (g/mol) 156.11
MDL Number MFCD01310985
InChI Key NXWTWYULZRDBSA-UHFFFAOYSA-N
Synonym 2-fluoro-4-hydroxy-benzoic acid, benzoic acid, 2-fluoro-4-hydroxy, pubchem2608, acmc-1b9jp, 4-hydroxyfluorobenzoic acid, frinton fr-2445, ksc352q5p, 2-fluoro-4-hydroxybenzoicacid, 4-hydroxy-2-fluorobenzoic acid, 2-fluoro4-hydroxy-benzoic acid
PubChem CID 2783398
IUPAC Name 2-fluoro-4-hydroxybenzoic acid
SMILES OC(=O)C1=C(F)C=C(O)C=C1

Specifications

Melting Point 187.0°C to 188.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Formula Weight 156.11
Percent Purity 98%
Chemical Name or Material 2-Fluoro-4-hydroxybenzoic acid
compliance-icons
Product Identifier
  • 2-Fluoro-4-hydroxybenzoic acid, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.