missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-desfluoro Bicalutamide, TRC

Product Code. 30398513
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30398513 10 mg
30405128 50 mg
30408399 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30398513 Supplier Toronto Research Chemicals Supplier No. TRCF59085010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1159977-36-2
Molecular Formula C18 H14 F4 N2 O4 S
Molecular Weight (g/mol) 430.3734
Synonym Propanamide, N-[4-cyano-3-(trifluoromethyl)phenyl]-3-[(2-fluorophenyl)sulfonyl]-2-hydroxy-2-methyl-, N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(2-fluorophenyl)sulfonyl]-2-hydroxy-2-methylpropanamide
IUPAC Name N-[4-cyano-3-(trifluoromethyl)phenyl]-3-(2-fluorophenyl)sulfonyl-2-hydroxy-2-methylpropanamide
SMILES CC(O)(CS(=O)(=O)c1ccccc1F)C(=O)Nc2ccc(C#N)c(c2)C(F)(F)F

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H14F4N2O4S/c1-17(26,10-29(27,28)15-5-3-2-4-14(15)19)16(25)24-12-7-6-11(9-23)13(8-12)18(20,21)22/h2-8,26H,10H2,1H3,(H,24,25)
Formula Weight 430.061
Chemical Name or Material (2RS)-N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(2-fluorophenyl)sulfonyl]-2-hydroxy-2-methylpropanamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.