missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-(trifluoromethyl)benzylamine, 97%, Thermo Scientific™

Product Code. 11348805
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11348805

Brand: Thermo Scientific Alfa Aesar L12428.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Fluoro-3-(trifluoromethyl)benzylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Not miscible or difficult to mix in water.

Notes
It is sensitive to air. Keep tightly closed. Store in a cool dry place. Store under dry inert gas. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 239135-49-0
Molecular Formula C8H7F4N
Molecular Weight (g/mol) 193.15
MDL Number MFCD00061161
InChI Key PQUYJDPJHOSOTN-UHFFFAOYSA-N
Synonym 2-fluoro-3-trifluoromethyl benzylamine, 2-fluoro-3-trifluoromethylbenzylamine, 2-fluoro-3-trifluoromethyl phenyl methanamine, benzenemethanamine, 2-fluoro-3-trifluoromethyl, 2-fluoro-3-trifluoromethyl phenyl methylamine, 1-2-fluoro-3-trifluoromethyl phenyl methanamine, otf-byl-5f, acmc-209uw1, rarechem al bd 0451
PubChem CID 2737565
IUPAC Name [2-fluoro-3-(trifluoromethyl)phenyl]methanamine
SMILES NCC1=CC=CC(=C1F)C(F)(F)F

Specifications

Density 1.321
Boiling Point 193°C
Flash Point 83°C (181°F)
Refractive Index 1.4548
Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 193.15
Percent Purity 97%
Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)benzylamine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.