missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-(trifluoromethyl)benzoic acid, 98%

Product Code. 11409786
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409786 1 g 1g
11419786 5 g 5g
11319629 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409786 Supplier Thermo Scientific Alfa Aesar Supplier No. B20280.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 115029-22-6 | C8H3F4O2 | 207.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115029-22-6
Molecular Formula C8H3F4O2
Molecular Weight (g/mol) 207.10
MDL Number MFCD00040980
InChI Key XVEAMDNSCPPPCP-UHFFFAOYSA-M
Synonym 2-fluoro-3-trifluoromethyl benzoic acid, 2-fluoro-3-trifluoromethyl benzoicacid, alpha,alpha,alpha,2-tetrafluoro-m-toluic acid, 2-fluoro-3-trifluoromethyl-benzoic acid, benzoic acid, 2-fluoro-3-trifluoromethyl, 2-fluoro-3-trifluoromethylbenzioc acid, pubchem2741, acmc-1boz3, ksc489q1l, rarechem al bo 0625
PubChem CID 518002
IUPAC Name 2-fluoro-3-(trifluoromethyl)benzoic acid
SMILES [O-]C(=O)C1=CC=CC(=C1F)C(F)(F)F

Specifications

Melting Point 126°C to 128°C
Quantity 1 g
Formula Weight 208.11
Percent Purity 98%
Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)benzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.