missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-3-(trifluoromethyl)aniline, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 2-Fluoro-3-(trifluoromethyl)aniline |
| CAS | 123973-25-1 |
| Molecular Formula | C7H5F4N |
| Quantity | 5 g |
| Synonym | 2-Fluoro-3-(trifluoromethyl)-benzenamine, [2-Fluoro-3-(trifluoromethyl)phenyl]amine |
| InChI Formula | InChI=1S/C7H5F4N/c8-6-4(7(9,10)11)2-1-3-5(6)12/h1-3H,12H2 |
| SMILES | Nc1cccc(c1F)C(F)(F)F |
| Molecular Weight (g/mol) | 179.11 |
| Formula Weight | 179.0358 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction