missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-nitropyridine, 98%, Thermo Scientific Chemicals

Product Code. 15447768
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Fluoro-3-nitropyridine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-3-nitropyridine is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1480-87-1
Molecular Formula C5H3FN2O2
Molecular Weight (g/mol) 142.09
MDL Number MFCD03095068
InChI Key QDKIYDGHCFZBGC-UHFFFAOYSA-N
Synonym 2-fluoro-3-nitro-pyridine, pyridine, 2-fluoro-3-nitro, pubchem1115, acmc-1bwlp, 2-fluor-3-nitropyridine, 2-fluoro-3-nitropyridine, ksc174g2n
PubChem CID 2762817
IUPAC Name 2-fluoro-3-nitropyridine
SMILES [O-][N+](=O)C1=C(F)N=CC=C1

Specifications

Melting Point 18°C
Color Yellow
Boiling Point 110°C (10 mmHg)
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 142.09
Percent Purity 98%
Physical Form Low Melting Solid
Chemical Name or Material 2-Fluoro-3-nitropyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'