missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-nitropyridine, 98%

Product Code. 15457768
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15457768

Brand: Thermo Scientific Alfa Aesar H61260.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1480-87-1 | C5H3FN2O2 | 142.09 g/mol

2-Fluoro-3-nitropyridine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-3-nitropyridine is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1480-87-1
Molecular Formula C5H3FN2O2
Molecular Weight (g/mol) 142.09
MDL Number MFCD03095068
InChI Key QDKIYDGHCFZBGC-UHFFFAOYSA-N
Synonym 2-fluoro-3-nitro-pyridine, pyridine, 2-fluoro-3-nitro, pubchem1115, acmc-1bwlp, 2-fluor-3-nitropyridine, 2-fluoro-3-nitropyridine, ksc174g2n
PubChem CID 2762817
IUPAC Name 2-fluoro-3-nitropyridine
SMILES [O-][N+](=O)C1=C(F)N=CC=C1

Specifications

Melting Point 18°C
Color Yellow
Boiling Point 110°C (10 mmHg)
Quantity 100 g
Solubility Information Slightly soluble in water.
Formula Weight 142.09
Percent Purity 98%
Physical Form Low Melting Solid
Chemical Name or Material 2-Fluoro-3-nitropyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.