missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-methylaniline, 97%

Product Code. 15457648
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15457648 25 g 25g
15467648 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15457648 Supplier Thermo Scientific Alfa Aesar Supplier No. H61143.14
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1978-33-2 | C7H8FN | 125.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1978-33-2
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.15
MDL Number MFCD06410915
InChI Key WFZUBZAEFXETBF-UHFFFAOYSA-N
Synonym 3-amino-2-fluorotoluene, 2-fluoro-m-toluidine, 2-fluoro-3-methylbenzenamine, 3-amino-2-fluoro-toluene, 2-fluoro-3-methyl-aniline, 2-fluoro-3-methylphenylamine, 2-fluoro-3-aminotoluene, benzenamine, 2-fluoro-3-methyl, 2-fluoro-3-methyl-phenylamine, pubchem1536
PubChem CID 22734623
IUPAC Name 2-fluoro-3-methylaniline
SMILES CC1=C(F)C(N)=CC=C1

Specifications

Boiling Point 87°C (12 mmHg)
Refractive Index 1.536
Quantity 25 g
UN Number UN2810
Beilstein 1231988
Formula Weight 125.14
Percent Purity 98%
Chemical Name or Material 2-Fluoro-3-methylaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.