missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-methoxybenzaldehyde, 96%, Thermo Scientific™

Product Code. 10264914 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10264914 5 g 5g
10153054 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10264914 Supplier Thermo Scientific Acros Supplier No. 425830050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 103438-88-6
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD03411962
InChI Key LIHCOUDNHILORI-UHFFFAOYSA-N
Synonym 2-fluoro-3-methoxy benzaldehyde, 2-fluoro-3-formylanisole, benzaldehyde, 2-fluoro-3-methoxy, zlchem 868, pubchem4224, acmc-1bs4m, flouro-3-methoxybenzaldehyde, 2-fluoro-3-methoxy-benzaldehyde, benzaldehyde,2-fluoro-3-methoxy, 2-fluoro-3-methoxybenzaldehyde
PubChem CID 3774463
IUPAC Name 2-fluoro-3-methoxybenzaldehyde
SMILES COC1=CC=CC(C=O)=C1F

Specifications

Melting Point 47°C to 51°C
Flash Point >110°C
Assay Percent Range 95% min. (GC)
Packaging Glass Bottle
Linear Formula FC6H3(OCH3)CHO
Quantity 5 g
Formula Weight 154.14
Percent Purity 96%
Chemical Name or Material 2-Fluoro-3-methoxybenzaldehyde
compliance-icons
Product Identifier
  • 2-Fluoro-3-methoxybenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.