missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-iodopyridine, 97%, Thermo Scientific™

Product Code. 11489260
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489260 1 g 1g
11479260 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489260 Supplier Thermo Scientific Alfa Aesar Supplier No. H50146.03
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113975-22-7
Molecular Formula C5H3FIN
Molecular Weight (g/mol) 222.989
MDL Number MFCD03095287
InChI Key WCDCAXVNBOLWNO-UHFFFAOYSA-N
Synonym 2-fluoro-3-iodo-pyridine, pyridine, 2-fluoro-3-iodo, zlchem 409, 2-fluoro-3 iodopyridine, 2-fluoro-3-iodo-pyridin, 2-fluoro-3-iodopyridine, 2-fluoro-3-iodopyridine;, 2-fluoro-3-iodo pyridine, acmc-1c73w, ksc174s0n
PubChem CID 7009501
IUPAC Name 2-fluoro-3-iodopyridine
SMILES C1=CC(=C(N=C1)F)I

Specifications

Quantity 1 g
Sensitivity Light sensitive
Formula Weight 222.99
Percent Purity 97%
Chemical Name or Material 2-Fluoro-3-iodopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.