missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-iodopyridine, 97%, Thermo Scientific™

Product Code. 10346374
Change view
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10346374 Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10346374 Supplier Thermo Scientific Maybridge Supplier No. MO00359DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 113975-22-7
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C5H3FIN
MDL Number MFCD03095287
Quantity 1 g
Synonym 2-fluoro-3-iodo-pyridine, pyridine, 2-fluoro-3-iodo, zlchem 409, 2-fluoro-3 iodopyridine, 2-fluoro-3-iodo-pyridin, 2-fluoro-3-iodopyridine, 2-fluoro-3-iodopyridine;, 2-fluoro-3-iodo pyridine, acmc-1c73w, ksc174s0n
InChI Key WCDCAXVNBOLWNO-UHFFFAOYSA-N
SMILES C1=CC(=C(N=C1)F)I
IUPAC Name 2-fluoro-3-iodopyridine
Molecular Weight (g/mol) 222.989
PubChem CID 7009501
Formula Weight 222.98
Percent Purity 97%
Chemical Name or Material 2-Fluoro-3-iodopyridine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.