missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-iodopyridine, 95%, Thermo Scientific™

Product Code. 15342052 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15342052 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15342052 Supplier Thermo Scientific Acros Supplier No. 454860010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 113975-22-7
Melting Point 45.0°C to 47.5°C
Assay Percent Range 95%
Packaging Glass Bottle
Molecular Formula C5H3FIN
Quantity 1 g
Synonym 2-fluoro-3-iodo-pyridine, pyridine, 2-fluoro-3-iodo, zlchem 409, 2-fluoro-3 iodopyridine, 2-fluoro-3-iodo-pyridin, 2-fluoro-3-iodopyridine, 2-fluoro-3-iodopyridine;, 2-fluoro-3-iodo pyridine, acmc-1c73w, ksc174s0n
InChI Key WCDCAXVNBOLWNO-UHFFFAOYSA-N
SMILES C1=CC(=C(N=C1)F)I
IUPAC Name 2-fluoro-3-iodopyridine
Molecular Weight (g/mol) 222.98
PubChem CID 7009501
Formula Weight 222.98
Percent Purity 95%
Chemical Name or Material 2-Fluoro-3-iodopyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Fluoro-3-iodopyridine, 95%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.