missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3,4-methylenedioxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog Number p-7054404
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1429340-48-6
2-Fluoro-3,4-methylenedioxyphenylboronic acid pinacol ester
This item is not returnable. View return policy
1429340-48-6
2-Fluoro-3,4-methylenedioxyphenylboronic acid pinacol ester

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 266.07
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-Fluoro-3,4-methylenedioxybenzeneboronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'