missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Fluoro-2'-deoxycytidine, 99%

Product Code. 15476999
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15476999 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15476999 Supplier Thermo Scientific Chemicals Supplier No. J65632.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10212-20-1 | C9H12FN3O4 | 245.21 g/mol

2'-Fluoro-2'-deoxycytidine is a potent inhibitor of the subgenomic hepatitis C virus replicon in Huh-7 cells. 2'-Fluoro-2'-deoxycytidine has been shown to inhibit Borna Disease virus replication and spread.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Fluoro-2′-deoxycytidine is a potent inhibitor of the subgenomic hepatitis C virus replicon in Huh-7 cells. 2′-Fluoro-2′-deoxycytidine has been shown to inhibit Borna Disease virus replication and spread.
TRUSTED_SUSTAINABILITY

Specifications

CAS 10212-20-1
Color White
Molecular Formula C9H12FN3O4
MDL Number MFCD00057445
Quantity 1 g
Synonym 2'-deoxy-2'-fluorocytidine, 2'-fluoro-2'-deoxycytidine, 4-amino-1-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl pyrimidin-2-one, cytidine, 2'-deoxy-2'-fluoro, unii-lcy080jpy9, 2-deoxy-2-fluorocytidine, lcy080jpy9, 2'-fc, 2'-fluoro-d-cytidine
Solubility Information Soluble in DMSO,Methanol.
InChI Key NVZFZMCNALTPBY-XVFCMESISA-N
SMILES C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)F
IUPAC Name 4-amino-1-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
Molecular Weight (g/mol) 245.21
PubChem CID 101507
Formula Weight 245.21
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2'-Fluoro-2'-deoxycytidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.