missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-1-methylpyridinium p-toluenesulfonate, tech. 90%

Product Code. 11387536
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387536 1 g 1g
11397536 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387536 Supplier Thermo Scientific Alfa Aesar Supplier No. L11088.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 58086-67-2 | C13H14FNO3S | 283.317 g/mol

2-Fluoro-1-methylpyridinium p-toluenesulfonate Is a peptide coupling reagent. Also used in the synthesis of ? -ribonucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-1-methylpyridinium p-toluenesulfonate Is a peptide coupling reagent. Also used in the synthesis of ɑ -ribonucleotides.

Solubility
Soluble in water (hydrolyzes), DMSO, dichloromethane, and acetonitrile.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58086-67-2
Molecular Formula C13H14FNO3S
Molecular Weight (g/mol) 283.317
MDL Number MFCD00011983
InChI Key HQWDKLAIDBOLFE-UHFFFAOYSA-M
Synonym 2-fluoro-1-methylpyridinium p-toluenesulfonate, 2-fluoro-1-methylpyridin-1-ium 4-methylbenzenesulfonate, 2-fluoro-1-methylpyridinium, mukaiyama's reagent, 2-fluoro-1-methylpyridinium 4-toluenesulfonate, 2-fluoro-1-methylpyridinium p-toluenesulphonate, 2-fluoro-n-methylpyridinium toluene-4-sulfonate, 2-fluoro-1-methylpyridin-1-ium tosylate, 2-fluoro-n-methylpyridiniumtoluene-4-sulphonate, 2-fluoro-1-methyl-pyridin-1-ium; 4-methylbenzenesulfonate
PubChem CID 171631
IUPAC Name 2-fluoro-1-methylpyridin-1-ium;4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)[O-].C[N+]1=CC=CC=C1F

Specifications

Melting Point 128°C to 131°C
Quantity 1 g
Beilstein 4345357
Sensitivity Moisture sensitive
Solubility Information Soluble in water (hydrolyzes),DMSO,dichloromethane,and acetonitrile.
Formula Weight 283.32
Percent Purity 90%
Grade Technical
Chemical Name or Material 2-Fluoro-1-methylpyridinium p-toluenesulfonate
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.